Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Tuning Forks Set
SDPSCI_ED

Catalog No. S05527 Shop All 3B Scientific Products
Change view
Click to view available options
Frequency:
440 Hz
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency
S05527 440 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05527 Supplier 3B Scientific™ Supplier No. U10120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Soft striking hammer and two tuning weights

For experiments in acoustics

  • Pair of identical forks mounted on separate resonant chambers made of clear grained fir wood
  • Natural frequency, internal length:
    • c' = 256 Hz 300 mm
    • e' = 322 Hz 240 mm
    • g' = 384 Hz 190 mm
    • c″ 512 Hz 140 mm

Specifications

Product Type Kit, Tuning Forks
For Use With (Application) For experiments in acoustics
Includes Soft striking hammer and two tuning weight
Weight (English) 2.20 lb.
Weight (Metric) 1 kg
Frequency 440 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.