Learn More
3B Scientific™ Vacuum Experiment Plate and Bell Jar
SDPSCI_ED
Catalog No. p-3996838 Shop All 3B Scientific Products
Change view
Click to view available options
Product Type:
Bell Jar
Vacuum Experiment Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S05534 Bell Jar
S05535 Vacuum Experiment Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05534 Supplier 3B Scientific™ Supplier No. U218511
Only null left
Add to Cart
Add to Cart
Includes
Plate: Hose connection on pump-side, ventilation cock and vacuum-tight current feed with 4mm safety sockets, center core with M12 thread for attaching experimental equipment vacuum jar
Assemble your own vacuum chamber in your classroom.
  • Combine vacuum plate and separate bell jar to create vacuum conditions
  • Useful in the coarse and fine vacuum range
  • Metal experiment plate has sealing ring on a tripod
  • Glass vacuum bell jar has a secure grip knob and polished flange
Order Info
Order both plate and bell jar to create vacuum conditions. Plate: current feed-through, two-pole with 4mm safety sockets (red, black); power feed, two cables (red, black) approximately 1m long with 4mm safety plugs; vacuum connection, two hose nozzles outer dia., 8 and 12mm. Electrical limit specifications: maximum voltage, 48V; maximum amperage, 12A. Dimensions (Dia. x H): Plate: 25.0 x 9.0cm; bell jar: 19.0 x 22.0cm.
Specifications
For Use With (Application) Coarse and fine vacuum range
Product Type Bell Jar