Learn More
Selleck Chemical LLC 3BDO 10mg 890405-51-3 3-benzyl-5-((2-nitrophenoxy) methyl)-dihydrofuran-2(3H)-one
SDP
Supplier:  Selleck Chemical LLC S831710MG
Encompass_Preferred
3BDO, a butyrolactone derivative, could target FKBP1A and activate the mTOR signaling pathway. It inhibits autophagy in HUVECs. 3BDO inhibits oxLDL-induced apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8134