Learn More
Cayman Chemical 3-Bromotyrosine (trifluoroacetate salt), Purity: ≥95%, CAS: 2320428-62-2, 10 mg
SDP
Catalog No. 502986509
Encompass_Preferred
A product of protein oxidation; a marker of eosinophil peroxidase-induced protein oxidation in vitro and in vivo
Catalog No. 50-298-6509 Supplier Cayman Chemical Supplier No. 2260610 MG
A product of protein oxidation; a marker of eosinophil peroxidase-induced protein oxidation in vitro and in vivo