Learn More
Genscript Corporation 3C protease, His, Human
Catalog No. NC0844672
Encompass
Recombinant PreScission Protease (rPSP) from human rhinovirus (HRV 3C) is a recombinant restriction-grade protease. PSP is a highly purified recombinant 6xHis-fusion protein, which recognizes the same cleavage site as the native enzyme. It is used to cleave affinity tags from fusion proteins. The optimum recognition site for this enzyme is the sequence Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro(LEVLFQ/GP) and cleavage occurs between the Gln and Gly-Pro residues. The optimal temperature for cleavage is 4°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of Recombinant PreScission Protease, reaction time, or incubation temperature. It can be removed by Ni2+ affinity resin.
Catalog No. NC0844672 Supplier Genscript Corporation Supplier No. Z03092500
Recombinant PreScission Protease (rPSP) from human rhinovirus (HRV 3C) is a recombinant restriction-grade protease. PSP is a highly purified recombinant 6xHis-fusion protein, which recognizes the same cleavage site as the native enzyme. It is used to cleave affinity tags from fusion proteins. The optimum recognition site for this enzyme is the sequence Leu-Glu-Val-Leu-Phe-Gln/Gly-Pro(LEVLFQ/GP) and cleavage occurs between the Gln and Gly-Pro residues. The optimal temperature for cleavage is 4°C. It is recommended that the cleavage for each fusion protein be optimized by varying the amount of Recombinant PreScission Protease, reaction time, or incubation temperature. It can be removed by Ni2+ affinity resin.