Learn More
Amstat Industries Staticmaster Brush with Ionizing Cartridge - 3"
SDP
Catalog No. NC0852066
Encompass
The Staticmaster 3C500 brush incorporates a proprietary Alpha ionizing technology into a smaller 3" static removal brush. It is an ideal solution for a variety of industrial, photographic, and analytical equipment such as precision balances. Staticmaster anti-static brushes are an excellent way to remove dust and other debris away from vinyl records. This will help maximize the life of your LP's. It is recommended to replace the cartridge every 12 months to ensure optimal performance.Amstat guarantees made to order new devices to ensure our customers get the longest use out of the ionizers.
Catalog No. NC0852066 Supplier Amstat Industries Supplier No. 3C500
The Staticmaster 3C500 brush incorporates a proprietary Alpha ionizing technology into a smaller 3" static removal brush. It is an ideal solution for a variety of industrial, photographic, and analytical equipment such as precision balances. Staticmaster anti-static brushes are an excellent way to remove dust and other debris away from vinyl records. This will help maximize the life of your LP's. It is recommended to replace the cartridge every 12 months to ensure optimal performance.Amstat guarantees made to order new devices to ensure our customers get the longest use out of the ionizers.