Learn More
Apexbio Technology LLC 3CAI 28755-03-5 5mg
SDP
Catalog No. 502240838
Encompass_Preferred
3CAI (CAS 28755-03-5) is a small molecule allosteric inhibitor selectively targeting AKT serine/threonine kinases Acting via an ATP-noncompetitive mechanism 3CAI binds directly to AKT1 and AKT2 exhibiting an IC50 of 1 M and suppressing 60% of AKT1 kinase activity at this concentration In vitro 3CAI reduces the expression of downstream AKT targets such as mTOR and GSK3 and at 4 M induces growth inhibition and apoptosis in HCT116 and HCT29 colon cancer cells Oral administration at 30 mg/kg significantly inhibits tumor growth and AKT substrate expression in mouse xenograft models 3CAI is valuable for studying AKT signaling and its implications in cancer research
This item is not returnable. View return policy
Catalog No. 50-224-0838 Supplier Apexbio Technology LLC Supplier No. B46885
3CAI (CAS 28755-03-5) is a small molecule allosteric inhibitor selectively targeting AKT serine/threonine kinases Acting via an ATP-noncompetitive mechanism 3CAI binds directly to AKT1 and AKT2 exhibiting an IC50 of 1 M and suppressing 60% of AKT1 kinase activity at this concentration In vitro 3CAI reduces the expression of downstream AKT targets such as mTOR and GSK3 and at 4 M induces growth inhibition and apoptosis in HCT116 and HCT29 colon cancer cells Oral administration at 30 mg/kg significantly inhibits tumor growth and AKT substrate expression in mouse xenograft models 3CAI is valuable for studying AKT signaling and its implications in cancer research