Learn More
STEMCELL Technologies 3 cc Syringes, Size: 100 Units
SDP
Catalog No. NC1749676
Encompass
Ideal for accurate dispensing of methylcellulose-based medium (e.g. MethoCult™), when used with Blunt-End Needles (Catalog #28110) purchased from STEMCELL Technologies to ensure compatibility and accuracy.
This item is not returnable. View return policy
Catalog No. NC1749676 Supplier STEMCELL Technologies Supplier No. 28240
Ideal for accurate dispensing of methylcellulose-based medium (e.g. MethoCult™), when used with Blunt-End Needles (Catalog #28110) purchased from STEMCELL Technologies to ensure compatibility and accuracy.