Learn More
Terumo Medical Corporation 3cc Syringe with Luer Slip Tip
Catalog No. 23600024 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Volume (Metric):
3 mL
Catalog No. Volume (Metric)
23-600-024 3 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-600-024 Supplier Terumo Medical Corporation Supplier No. SS03S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Graduated syringe without needle
  • Luer slip tip
  • Graduations: 0.1cc (3cc total)
  • 100 / box, 10 boxes / case (1,000)
Specifications
Material Polypropylene (Barrel), Polypropylene (Plunger)
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe without needle
Disposable Single-use
Graduations 0.1 cc
Volume (Metric) 3 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Slip
Graduated Yes
Sterility Sterile