Learn More
Sigma Aldrich Fine Chemicals Biosciences 3CL Mpro protease fluoroge1MG
Catalog No. 502906068
Encompass
This is substrate peptide for Main protease from SARS-Cov2 3CL protease (Mpro) . This substrate allows measuring the activity of 3CL protease using a standard plate reader or spectrophotometer.Proteolytic cleavage by 3CL protease will release the flourecent AfC group from the peptide. After relase an increase in floursence is obtained (Ex 400nm Em 505nm) .Thr-Ser-Ala-Val-Leu-Gln-AfCLyophilized from 0.1% TfA in H2O
Catalog No. 50-290-6068 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE01801MG
This is substrate peptide for Main protease from SARS-Cov2 3CL protease (Mpro) . This substrate allows measuring the activity of 3CL protease using a standard plate reader or spectrophotometer.Proteolytic cleavage by 3CL protease will release the flourecent AfC group from the peptide. After relase an increase in floursence is obtained (Ex 400nm Em 505nm) .Thr-Ser-Ala-Val-Leu-Gln-AfCLyophilized from 0.1% TfA in H2O