Learn More
Perkin Elmer US LLC Brownlee Pecosphere RA C18 Cartridge Column - 33 mm x 4.6 mm I.D., Pkg. 5
Catalog No. NC9164520
Encompass_Preferred
Brownlee Pecosphere RA C18 columns are packed with rugged, high purity silica with standard end-capping for the analysis of basic analytes. These cartridge-style columns require the 33 mm fast LC Pecosphere cartridge holder (07150028). Quantity: Package of 5 End-Capping: Yes Carbon Load: 12% Interaction Mode: Reversed Phase pH Range: 2.0-8.0 Surface Area: 200 m2/g USP Code: L1
Catalog No. NC9164520 Supplier Perkin Elmer US LLC Supplier No. 02580195
Brownlee Pecosphere RA C18 columns are packed with rugged, high purity silica with standard end-capping for the analysis of basic analytes. These cartridge-style columns require the 33 mm fast LC Pecosphere cartridge holder (07150028). Quantity: Package of 5 End-Capping: Yes Carbon Load: 12% Interaction Mode: Reversed Phase pH Range: 2.0-8.0 Surface Area: 200 m2/g USP Code: L1