Learn More
eMolecules​ 3-Cyano-4-oxo-piperidine-1-carboxylic acid tert-butyl ester | Combi-Blocks | 914988-10-6 | MFCD11109814 | 224.260 | C11H16N2O3 | 97.000 | CC(C)(C)OC(=O)N1CCC(=O)C(C1)C#N | 10g | 448006621
Supplier:  eMolecules​ 448006621
Encompass_Preferred
3-Cyano-4-oxo-piperidine-1-carboxylic acid tert-butyl ester | Combi-Blocks | 914988-10-6 | MFCD11109814 | 224.260 | C11H16N2O3 | 97.000 | CC(C)(C)OC(=O)N1CCC(=O)C(C1)C#N | 10g | 448006621
Catalog No. NC2005119