Learn More
Cayman Chemical 3-Cysteinylacetaminophen (trifluoroacetate salt), Purity: ≥98%, CAS: 1331891-93-0, 10 mg
SDP
Catalog No. 502933762
Encompass_Preferred
An acetaminophen-protein adduct; has been found in isolated human serum following therapeutic and supratherapeutic doses of acetaminophen and in the presence and absence of hepatotoxicity; decreases renal GSH levels
Catalog No. 50-293-3762 Supplier Cayman Chemical Supplier No. 2638810 MG
An acetaminophen-protein adduct; has been found in isolated human serum following therapeutic and supratherapeutic doses of acetaminophen and in the presence and absence of hepatotoxicity; decreases renal GSH levels