Learn More
Marketlab 3D/4D Eclipse Endocavity Probe Cover
Catalog No. 22505341
Change view
Click to view available options
Length (English):
9.5 in.
Catalog No. Length (English)
22-505-341 9.5 in.
1 options
Catalog No. 22-505-341 Supplier Marketlab Supplier No. 911447
Add to Cart
Add to Cart
The Eclipse Probe Cover is a disposable probe cover, pre-gelled inside with AQUASONIC™ 100 Ultrasound Transmission Gel.
  • Made of pliable, yet strong polyethylene film, reducing the concerns of breakage, and thereby protecting the probe/transducer during ultrasound procedures.
  • Endocavity
  • Non-sterile
  • Latex-free
Specifications
Quantity 100/Pk.
Length (English) 9.5 in.