Learn More
Alkali Scientific BenchWaver™ 3D Orbital Shaker with Non-slip Mat for Western Blot, 115V
SDP
Catalog No. 502389319
Encompass_Preferred
The BenchWaver™ 3D combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional "undulating" motion commonly preferred for molecular biology applications. The tilt angle is instantly adjustable (no tools or disassembly required) from 0 to 10° and the mixing speed can be adjusted from 5 to 105 rpm, using the large, user-friendly LCD control panel. Combined control of both parameters (tilt and speed) makes the BenchWaver a versatile workhorse that can provide anything from gentle, slow moving waves to a rapid, more vigorous motion. Additionally, time of operation can be selected from 1 minute up to 99 hours or for continuous operation. The large 13.5 x 13 inch platform is supplied complete with a non-slip, rubber mat. BenchWaver™ is a reliable and versatile instrument and can be used in refrigerated or incubated environments from 2° to 60°C.
Catalog No. 50-238-9319 Supplier Alkali Scientific Supplier No. RS3500
The BenchWaver™ 3D combines the basic rocking motion of traditional platform rockers with the circular motion of an orbital shaker. This combination is ideal for generating the optimum 3-dimensional "undulating" motion commonly preferred for molecular biology applications. The tilt angle is instantly adjustable (no tools or disassembly required) from 0 to 10° and the mixing speed can be adjusted from 5 to 105 rpm, using the large, user-friendly LCD control panel. Combined control of both parameters (tilt and speed) makes the BenchWaver a versatile workhorse that can provide anything from gentle, slow moving waves to a rapid, more vigorous motion. Additionally, time of operation can be selected from 1 minute up to 99 hours or for continuous operation. The large 13.5 x 13 inch platform is supplied complete with a non-slip, rubber mat. BenchWaver™ is a reliable and versatile instrument and can be used in refrigerated or incubated environments from 2° to 60°C.