Learn More
S BIO PrimeSurface 3D culture: Ultra-low Attachment 90 mm dish
Catalog No. NC1713414
Encompass
PrimeSurface cultureware consists of ultra-low attachment (ULA) dishes are designed to promote scaffold-free, self-assembly spheroid formation with uniform size and shape. These dishes are pre-coated with a unique ultra-hydrophilic polymer that facilitates spontaneous spheroid formation. Their high optical clarity makes them highly suitable for bright-field imaging and confocal microscopy.
Catalog No. NC1713414 Supplier S BIO Supplier No. MS90900
PrimeSurface cultureware consists of ultra-low attachment (ULA) dishes are designed to promote scaffold-free, self-assembly spheroid formation with uniform size and shape. These dishes are pre-coated with a unique ultra-hydrophilic polymer that facilitates spontaneous spheroid formation. Their high optical clarity makes them highly suitable for bright-field imaging and confocal microscopy.