Learn More
Sciencell Research Laboratories Ready-to-use 3D Human Cortical Spheroids, 48
Catalog No. NC2610513
Encompass
ScienCell's ready-to-use 3D human cortical spheroids (SP3D-HCS) are comprised of primary human neurons and astrocytes at a 1:7 coculturing ratio. SP3D-HCS maintain direct cell-cell interactions between neurons and astrocytes and form functional synapses throughout the spheroids. This advanced and ready-to-use cortical spheroid is highly suitable for the study of CNS functions, diseases, and therapeutics.
Catalog No. NC2610513 Supplier Sciencell Research Laboratories Supplier No. SP3D152048WELL
ScienCell's ready-to-use 3D human cortical spheroids (SP3D-HCS) are comprised of primary human neurons and astrocytes at a 1:7 coculturing ratio. SP3D-HCS maintain direct cell-cell interactions between neurons and astrocytes and form functional synapses throughout the spheroids. This advanced and ready-to-use cortical spheroid is highly suitable for the study of CNS functions, diseases, and therapeutics.