Learn More
ILEX LIFE SCIENCES LLC 3D LIFE 10X CBBUFF PH7.2 600UL
SDP
Catalog No. 502733954
Encompass_Preferred
3-D Life 10x CB Buffer (pH 7.2), buffer component for slow-gelling 3-D Life Hydrogels (600 ul). INTRO: 3-D Life technology, manufactured in Germany by Cellendes GmbH, is an integrated system for the generation of biomimetic hydrogels used in 3D cell culture. 3-D Life represents the most flexible system for biomimetic hydrogel design. DESCRIPTION: 3-D Life 10x CB-Buffer (pH 7.2) is a 10x concentrated salt and buffer solution which is diluted to physiological strength when used in the 3-D Life Hydrogel system. It is used for the formation of slow gelling 3-D Life Hydrogels where it provides a cell compatible environment at pH 7.2. In conjunction with 3-D Life 10x CB-Buffer (pH 5.5) it is used to generate pHs of intermediate values to modify the gelation kinetics of fast gelling gels. APPLICATION: Used with other components of the 3-D Life Hydrogel System to establish 3-D cell cultures. QUANTITY: 600 µl. Sufficient for at least 6 ml 3-D Life Hydrogel.
Catalog No. 50-273-3954 Supplier ILEX LIFE SCIENCES LLC Supplier No. B203
3-D Life 10x CB Buffer (pH 7.2), buffer component for slow-gelling 3-D Life Hydrogels (600 ul). INTRO: 3-D Life technology, manufactured in Germany by Cellendes GmbH, is an integrated system for the generation of biomimetic hydrogels used in 3D cell culture. 3-D Life represents the most flexible system for biomimetic hydrogel design. DESCRIPTION: 3-D Life 10x CB-Buffer (pH 7.2) is a 10x concentrated salt and buffer solution which is diluted to physiological strength when used in the 3-D Life Hydrogel system. It is used for the formation of slow gelling 3-D Life Hydrogels where it provides a cell compatible environment at pH 7.2. In conjunction with 3-D Life 10x CB-Buffer (pH 5.5) it is used to generate pHs of intermediate values to modify the gelation kinetics of fast gelling gels. APPLICATION: Used with other components of the 3-D Life Hydrogel System to establish 3-D cell cultures. QUANTITY: 600 µl. Sufficient for at least 6 ml 3-D Life Hydrogel.