Learn More
ILEX LIFE SCIENCES LLC Cellendes 3-D Life Dextranase, Hydrogel Accessory (500 ul)
SDP
Supplier:  ILEX LIFE SCIENCES LLC D101
Encompass_Preferred
3-D Life is the most flexible system for biomimetic hydrogel design for 3D cell cultures. It uses two polymers that form a hydrogel by covalent crosslinking at ambient temperature. The 3-D Life Dextranase enzyme allows the degradation of dextran-based 3-D Life Hydrogels. It contains dextranase from Chaetomium sp., which cleaves glycosidic bonds of dextran. Application: Dissolution of dextran-based hydrogels to recover live or fixed cells without proteolytic degradation. 3-D Life Dextranase is used at a 1:20 dilution in the culture supernatant to dissolve dextran-based hydrogels. A detailed description of application is available in the 3-D Life Hydrogels User Guide. Concentration of reactive groups: greater than 4000 units/mL. Manufactured in Germany by Cellendes GmbH.
Catalog No. NC2081790