Learn More
JDM LAB SOLUTIONS LLC 3D Mini Roller
Catalog No. 5000014324
Encompass
3D Mini Roller (Stackable) MR-03UA Features Variable speed and LED display allow for precise control Anti-corrosive and wear-resistant construction is safe for high humidity and CO2 environments Easily add or remove rollers to fit a wide range of tube and bottle sizes Stackable design allows for efficient use of benchtop surface area Included wired remote externally control instrument when running inside the incubator
Catalog No. 50-000-14324 Supplier JDM LAB SOLUTIONS LLC Supplier No. MR03UA
3D Mini Roller (Stackable) MR-03UA Features Variable speed and LED display allow for precise control Anti-corrosive and wear-resistant construction is safe for high humidity and CO2 environments Easily add or remove rollers to fit a wide range of tube and bottle sizes Stackable design allows for efficient use of benchtop surface area Included wired remote externally control instrument when running inside the incubator