Learn More
Grainger Scientific Industries 3D Orbital Shaker Attachment, Metal, For Enviro-Genie and Incubator-Genie, 1 ea
Catalog No. 502768716
Encompass_Preferred
This 3-D orbital shaker attachment is designed to expand the functionality of compatible shakers, allowing for complex motion profiles in laboratory applications. It provides a specialized platform movement, enabling both rocking and rotating actions for effective sample mixing.
  • Provides a 5° angle through a 360° rotation for comprehensive agitation.
  • Includes a non-slip mat to secure vessels during operation.
  • Compatible with Enviro-Genie and Incubator-Genie units only.
  • Constructed from durable metal for reliable laboratory use.
  • Features a platform capacity of 4.5 kg.
  • Suitable for use in cold rooms and incubators within a temperature range of 0°C to 38°C.
Catalog No. 50-276-8716 Supplier Grainger Supplier No. 13R360
This 3-D orbital shaker attachment is designed to expand the functionality of compatible shakers, allowing for complex motion profiles in laboratory applications. It provides a specialized platform movement, enabling both rocking and rotating actions for effective sample mixing.
  • Provides a 5° angle through a 360° rotation for comprehensive agitation.
  • Includes a non-slip mat to secure vessels during operation.
  • Compatible with Enviro-Genie and Incubator-Genie units only.
  • Constructed from durable metal for reliable laboratory use.
  • Features a platform capacity of 4.5 kg.
  • Suitable for use in cold rooms and incubators within a temperature range of 0°C to 38°C.