Learn More
McMaster-Carr 3D-Printed Part Removal Tool, 4 1/4"L
Catalog No. NC1765432
Encompass
Separate 3D-printed parts from 3D-printer plates without damaging either one of them. Unlike conventional putty knives or spatulas, these tools have a right-angle handle and tapered blade, which makes it easier to slide them under parts and loosen them from the plates. The blade’s rounded edge prevents nicks and scrapes. The rubber handle gives you a nonslip grip.
Catalog No. NC1765432 Supplier McMaster-Carr Supplier No. 3518N11
Separate 3D-printed parts from 3D-printer plates without damaging either one of them. Unlike conventional putty knives or spatulas, these tools have a right-angle handle and tapered blade, which makes it easier to slide them under parts and loosen them from the plates. The blade’s rounded edge prevents nicks and scrapes. The rubber handle gives you a nonslip grip.