Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TerraCycle™ 3D Printing Materials Zero Waste Box™
SDPSCI_ED

Catalog No. S88854135
Change view
Click to view available options
Capacity (Metric):
39.66 L
79.31 L
136.42 L
Dimensions (L x W x H):
11 x 11 x 20 in.
11 x 11 x 40 in.
15 x 15 x 37 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Dimensions (L x W x H)
S88854135 39.66 L 11 x 11 x 20 in.
S88854136 79.31 L 11 x 11 x 40 in.
S88854137 136.42 L 15 x 15 x 37 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options

Not available in Hawaii, Alaska, and Puerto Rico.

Catalog No. S88854135 Supplier TerraCycle™ Supplier No. CS119S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Not available in Hawaii, Alaska, and Puerto Rico.

Recycle 3D Printing Materials. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.

  • WHAT YOU CAN RECYCLE:
    • Accepted Waste: Please send us any printing spools and scrap printing material generated by your 3D printer including ABS, HIPS, Nylon, and TPU.
    • Unaccepted Waste: Do not send in 3D printers, 3D printer parts, traditional printer cartridges, or 3D prints of any material other than ABS, HIPS, Nylon, and TPU. Printing materials made from PP, PE, PS, and PTEG may be collected in our Plastic Packaging Zero Waste Box. Printing Materials made from PLA may be collected in our Biodegradable Plastics Zero Waste Box.
  • How It Works
    • Order: Select a Zero Waste Box system and place your order.
    • Collect: Collect the accepted items to fill the system.
    • Ship: Send it back to TerraCycle with the prepaid return label on your system.*
    • Reorder: Order your next Zero Waste Box system to continue to recycle everything.
  • What’s Included?
    • One Zero Waste Box
    • One prepaid return label, affixed to your box
    • Processing and recycling of the materials
    • Customer support from our team of experts
  • Important Shipping Restrictions
    • For the Zero Waste Box system to work effectively and safely, please follow these simple shipping guidelines on how and what you can ship back to TerraCycle:
      • Only send in items this system accepts. Sending in items that fall outside the selected category causes contamination and the need for added separation, which increases costs.
      • Do not send in household items that may be considered hazardous waste. These include items such as lithium-ion batteries, pressurized canisters, pesticides, oil-based paint or medical sharps, organics, broken glass, biomedical waste, soiled diapers, or expired medication. Do not send in anything sharp, flammable, reactive, corrosive, ignitable, radioactive, toxic, infectious, or pathogenic.
      • Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
      • Do not ship dripping packages. Please make sure all excess product has been removed before putting it in your Zero Waste Box, Pouch, or Pallet. If you choose to rinse your items, please note that the system must be completely dry prior to shipping.

Specifications

Capacity (Metric) 39.66 L
Capacity (English) 39.66 L
Color Brown, Green, White
Dimensions (L x W x H) 11 x 11 x 20 in.
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Autoclavable Not Autoclavable
Includes Cardboard Box
Liner Material Plastic
For Use With (Application) Recycling
Width (English) 11 in.
Length (English) 11 in.
Thickness 11 in.
Height (English) 20 in.
Material Cardboard
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.