Learn More
Promocell GmbH PromoCell 3D Tumorsphere Medium XF prf 250 ml, with C-39870 attached to flask
Catalog No. NC2004523
Encompass
The PromoCell 3D Tumorsphere Medium XF is a culture system developed for the standardized serial culture of cancer cell lines as tumorspheres/mammospheres. The formulation supports sustained cell proliferation allowing for serial passage of the 3D culture. Its serum-free and xeno-free formulation provides a culture environment devoid of all stimuli originating from non-defined materials.Components:The PromoCell 3D Tumorsphere Medium XF consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete medium.Key features:Establish 3D tumorsphere cultures directly from 2D cancer cell lines Suitable for long-term routine culture of tumorspheres Compatible with most commonly used cancer cell lines Xeno-free and serum-free formulation
Catalog No. NC2004523 Supplier Promocell GmbH Supplier No. C28075
The PromoCell 3D Tumorsphere Medium XF is a culture system developed for the standardized serial culture of cancer cell lines as tumorspheres/mammospheres. The formulation supports sustained cell proliferation allowing for serial passage of the 3D culture. Its serum-free and xeno-free formulation provides a culture environment devoid of all stimuli originating from non-defined materials.Components:The PromoCell 3D Tumorsphere Medium XF consists of a bottle of Basal Medium and one vial of SupplementMix. Adding the SupplementMix to the Basal Medium results in the complete medium.Key features:Establish 3D tumorsphere cultures directly from 2D cancer cell lines Suitable for long-term routine culture of tumorspheres Compatible with most commonly used cancer cell lines Xeno-free and serum-free formulation