Learn More
ATCC 3D4/31; Monomyeloid Cell; Pig (Sus scrofa)
Catalog No. 502384715
Encompass
3D4/31 is an alveolar cell line that was isolated in 1998 from the lung of a 27-day-old pig. This cell line was deposited by J Gren and can be used in immunology research.
Catalog No. 50-238-4715 Supplier ATCC Supplier No. CRL2844
3D4/31 is an alveolar cell line that was isolated in 1998 from the lung of a 27-day-old pig. This cell line was deposited by J Gren and can be used in immunology research.