Learn More
Cayman Chemical 3DeazaadenosInhydrochLRI 10mg
SDP
Catalog No. 502988361
Encompass_Preferred
An inhibitor of S-adenosylhomocysteine hydrolase (Ki = 3.9 μM); has anti-inflammatory properties, inhibiting leukocyte adhesion and chemotaxis, lymphocyte-mediated cytolysis, phagocytosis, degranulation, and NF-κB signaling; has antiviral and antibacterial activities
Catalog No. 50-298-8361 Supplier Cayman Chemical Supplier No. 3096910 MG
An inhibitor of S-adenosylhomocysteine hydrolase (Ki = 3.9 μM); has anti-inflammatory properties, inhibiting leukocyte adhesion and chemotaxis, lymphocyte-mediated cytolysis, phagocytosis, degranulation, and NF-κB signaling; has antiviral and antibacterial activities