Learn More
Cayman Chemical 3-Deoxy-D-glycero-D-galacto-2-nonulosonic Acid, Purity: ≥95%, CAS: 153666-19-4, 10 mg
SDP
Catalog No. 502967172
Encompass_Preferred
A naturally occuring deaminated sialic acid that is thought to be a precursor for the biosynthesis of other members of the sialic acid family; can be used to analyze nonulosonic acid residues in polysialoglycoproteins
Catalog No. 50-296-7172 Supplier Cayman Chemical Supplier No. 1786210 MG
A naturally occuring deaminated sialic acid that is thought to be a precursor for the biosynthesis of other members of the sialic acid family; can be used to analyze nonulosonic acid residues in polysialoglycoproteins