Learn More
Description
- Uses organotypic 3D cultures during the medium-air incubation step
- Media produces complex stratified squamous epithelium containing both proliferating (BrdU+) and cornified (Filaggrin+) cell polulations within 10 days
- Media is complete and does not require further supplementation
- No growth
Specifications
Specifications
| Content And Storage | Store -20°C, After thawing store 4°C, Use in two weeks |
| Product Type | Skin Differentiation Medium |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Endotoxin Level | <2 EU/mL |
| For Use With (Application) | Cell Culture, Cell Differentiation, Cosmetics |
| Osmolality | 326 to 360 |
| pH | 7.0 to 7.6 |
| Form | Liquid |
For research purposes only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.