Learn More
Epredia™ 3DHistech 3D View Software - 3 Users
Catalog No. 91186 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Microarray Scanner
Catalog No. For Use With (Equipment)
91-186 Microarray Scanner
1 options
Catalog No. 91-186 Supplier Epredia™ Supplier No. 91186
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech 3D View is a software application that prepares 3D reconstructions of 2D serial sections. It is also capable of displaying microCT images in 3D (VOL/VGI format).
While microscope slides allow you to see only one section of reality, 3DView is a tool that can reconstruct the original tissue from its serial sections. The 3D image can then be rotated, zoomed into and 'sliced' according to user needs.
How 3DView Works
  • Aligns several 2D serial sections using the SlideMatch function
  • Prepares a 3D reconstruction from 2D serial sections
  • The 3D image can then be rotated, zoomed into and 'sliced' according to user needs
Key features
  • Advanced visualization options
  • Measurement tools to assess metric distances
  • Advanced file export options
  • Wide compatibility with third-party file formats
  • Minimum system requirements
  • License for 3 users - Research use only
Specifications
Product Type Software
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.