Learn More
Epredia™ 3DHistech CellQuant Cell Detection Module for IHC Stained Samples - 3 Users
Catalog No. 91242 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Microarray Scanner
Catalog No. For Use With (Equipment)
91-242 Microarray Scanner
1 options
Catalog No. 91-242 Supplier Epredia™ Supplier No. 91242
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech CellQuant cell detection module is a versatile cell detection application, an optimal solution for investigating various IHC stainings (best suited for Ki67 slides).
  • A special IHC color calibration option is integrated into these quantification modules, facilitating software integration to the local laboratory
  • By applying this function the software can be calibrated to the sample quality (local laboratory stain protocol, or different stainer), resulting in an adequate measurement
  • The Swiss army knife of 3DHISTECH's image analysis tools, CellQuant is a versatile cell detection application, an optimal solution for investigating various IHC stainings (best suited for Ki67 slides)
  • The application is adequate for cell nuclei, cytoplasmic and membrane marker quantification
  • The software reports the results like positivity ranges of cell nuclei, cytoplasm or membrane signals
  • This module can analyze fluorescent and brightfield slides
  • License for 3 users - Research use only
Specifications
Product Type Cell Detection Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.