Learn More
Epredia™ 3DHistech CellQuant Cell Detection Module for IHC Stained Samples - 5 Users
Shop All Epredia ProductsDescription
- A special IHC color calibration option is integrated into these quantification modules, facilitating software integration to the local laboratory
- By applying this function the software can be calibrated to the sample quality (local laboratory stain protocol, or different stainer), resulting in an adequate measurement
- The Swiss army knife of 3DHISTECH's image analysis tools, CellQuant is a versatile cell detection application, an optimal solution for investigating various IHC stainings (best suited for Ki67 slides)
- The application is adequate for cell nuclei, cytoplasmic and membrane marker quantification
- The software reports the results like positivity ranges of cell nuclei, cytoplasm or membrane signals
- This module can analyze fluorescent and brightfield slides
- License for 5 users - Research use only
Specifications
Specifications
| Product Type | Cell Detection Module |
| For Use With (Application) | Digital Pathology |
| For Use With (Equipment) | Microarray Scanner |
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.