Learn More
Epredia™ 3DHistech CISHQuant and CISH-RNAQuant Image Analysis Modules - 10 Users
Catalog No. 91234 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Microarray Scanner
Catalog No. For Use With (Equipment)
91-234 Microarray Scanner
1 options
Catalog No. 91-234 Supplier Epredia™ Supplier No. 91234
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech CISHQuant and CISH-RNAQuant image analysis quantify CISH (Chromogenic In Situ Hybridization) stained samples.
  • CISHQuant quantifies CISH (Chromogenic In Situ Hybridization) stained brightfield samples
  • The algorithm can be calibrated to the staining protocol and quality by using an integrated colour setting tool
  • This module is suitable for examining genetic mutations (gene amplification, deletion) and chromosomal aberrations (polysomy, aneusomy) on CISH slides
  • License for 10 users - Research use only
Specifications
Product Type Image Analysis Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.