Learn More
Epredia™ 3DHistech DensitoQuant Stain-Intensity-Based IHC Quantification Module - 5 Users
Catalog No. 91093 Shop All Epredia Products
Change view
Click to view available options
5 Users
Catalog No.
91-093 5 Users
1 options
Catalog No. 91-093 Supplier Epredia™ Supplier No. 91093
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech DensitoQuant is an easy-to-use, fast and accurate, stain-intensity-based IHC quantification tool.
  • A special IHC color calibration option is integrated into these quantification modules, facilitating software integration to the local laboratory. By applying this function the software can be calibrated to the sample quality (local laboratory stain protocol, or different stainer), resulting in an adequate measurement.
  • DensitoQuant is an easy-to-use, fast and accurate, stain-intensity-based IHC quantification tool. The application identifies the positive stain, based on an automatic color separation method through which individual positive pixels are counted and classified based on intensity and threshold ranges. This way, an H-Score can be calculated based on the proportion of positive and negative pixels.
  • License for 5 users - Research use only
Specifications
Product Type IHC Quantification Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.