Learn More
Epredia™ 3DHistech Fluorescence Image Analysis Module - 5 Users
Catalog No. RAS91153 Shop All Epredia Products
Change view
Click to view available options
5 Users
Catalog No.
RAS91153 5 Users
1 options
Catalog No. RAS91153 Supplier Epredia™ Supplier No. 91153
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Fluorescence quantification package includes HistoQuant and FISHQuant.
  • HistoQuant: an image segmentation module which identifies stained tissue elements based on color and intensity features
  • FISHQuant: a powerful cancer and cytogenetic application dedicated to quantifying FISH (Fluorescent In Situ Hybridization) signals on tissue samples of solid tumour diseases like breast cancer, lung cancer, sarcoma symptoms, and lymphomas
  • License for 5 users - Research use only
Specifications
Product Type Image Analysis Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.