Learn More
Epredia™ 3DHistech HistoQuant Image Segmentation Module - 1 User
Catalog No. 91081 Shop All Epredia Products
Change view
Click to view available options
1 User
Catalog No.
91-081 1 User
1 options
Catalog No. 91-081 Supplier Epredia™ Supplier No. 91081
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech HistoQuant is an image segmentation module which identifies stained tissue elements based on color and intensity features.
  • Color- and pattern-based image analysis solutions are used to identify different tissue elements in stained tissue. Instead of the time-consuming manual annotation, automated detection and annotation of a region of interest is available with a quick training on relevant tissue samples.
  • HistoQuant is an image segmentation module which identifies stained tissue elements based on color and intensity features. This is an adequate solution for double-stain quantification or multiplex fluorescent analysis, with dual stain colocalization option. This module can analyze fluorescent and brightfield slides.
  • License for 1 user - Research use only
Specifications
Product Type Image Segmentation Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.