Learn More
Epredia™ 3DHistech IHC Image Analysis Module - 5 Users
Catalog No. 91143 Shop All Epredia Products
Change view
Click to view available options
5 Users
Catalog No.
91-143 5 Users
1 options
Catalog No. 91-143 Supplier Epredia™ Supplier No. 91143
Add to Cart
Add to Cart
Epredia™ 3DHistech IHC quantification software package includes PatternQuant, NuclearQuant, MembraneQuant and DensitoQuant.
  • PatternQuant: a trainable pattern recognition module for tissue classification, tissue pre-segmentation and identification of several tissue structures
  • NuclearQuant: designed for cell nuclei detection and quantification of IHC stained samples
  • MembraneQuant: a cell membrane detection software application that can be used for the quantification of IHC stained histological samples
  • DensitoQuant: an easy-to-use, fast and accurate, stain-intensity-based IHC quantification tool
  • License for 5 users - Research use only
Specifications
Product Type Image Analysis Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.