Learn More
Epredia™ 3DHistech MembraneQuant Cell Membrane Detection Module - 10 Users
Catalog No. 91044 Shop All Epredia Products
Change view
Click to view available options
10 Users
Catalog No.
91-044 10 Users
1 options
Catalog No. 91-044 Supplier Epredia™ Supplier No. 91044
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech MembraneQuant is a cell membrane detection software application that can be used for the quantification of IHC stained histological samples.
  • A special IHC color calibration option is integrated into these quantification modules, facilitating software integration to the local laboratory. By applying this function the software can be calibrated to the sample quality (local laboratory stain protocol, or different stainer), resulting in an adequate measurement.
  • MembraneQuant is a cell membrane detection software application that can be used for the quantification of IHC stained histological samples. This module measures cell morphology and stain density, reports intensity-based core ranges, overall scores and positivity percentages (including H-Score), distinguishing the continuous membrane stain from the moderate stain.
  • License for 10 users - Research use only
Specifications
Product Type Cell Membrane Detection Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.