Learn More
Epredia™ 3DHistech NuclearQuant Cell Nuclei Detection Module - 1 User
Catalog No. 91031 Shop All Epredia Products
Change view
Click to view available options
1 User
Catalog No.
91-031 1 User
1 options
Catalog No. 91-031 Supplier Epredia™ Supplier No. 91031
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech NuclearQuant is designed for cell nuclei detection and quantification of IHC stained samples.
  • A special IHC color calibration option is integrated into these quantification modules, facilitating software integration to the local laboratory. By applying this function the software can be calibrated to the sample quality (local laboratory stain protocol, or different stainer), resulting in an adequate measurement.
  • NuclearQuant is designed for cell nuclei detection and quantification of IHC stained samples. NuclearQuant measures several morphological features besides stain intensity. The cell nuclei classification and the final score are calculated by the intensity score and the proportion score.
  • License for 1 user - Research use only
Specifications
Product Type Cell Nuclei Detection Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.