Learn More
Epredia™ 3DHistech Pannoramic 250 Flash III Digital Scanner, Brightfield to 80x Resolution
Catalog No. F3BF4080S Shop All Epredia Products
Change view
Click to view available options
Resolution:
Brightfield to 80x
Catalog No. Resolution
F3BF4080S Brightfield to 80x
1 options
Catalog No. F3BF4080S Supplier Epredia™ Supplier No. F3BF4080S
Add to Cart
Add to Cart
Epredia™ 3DHistech Pannoramic 250 Flash III whole-slide digital scanner is an all-in-one solution for labs that demand BF and/or FL scanning on the same platform with award winning quality and continuous loading of up to 300 slides at FLASH technology speeds.
  • 300-slide capacity and continuous loading with vertical slide arrangement
  • Award-winning, exceptional image quality for brightfield
  • Pulsed Xenon FLASH light source for high-speed brightfield scanning
  • Up to 80x brightfield
  • Motorized objective and camera changer
  • Automatic slide loading, previewing, barcode reading and scanning
  • All-around system for high volume slide scanning - Research use only
Specifications
Resolution Brightfield to 80x
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.