Learn More
Epredia™ 3DHistech Pannoramic 480 Digital Scanner, Brightfield to 80x Resolution
Catalog No. P4804080S Shop All Epredia Products
Change view
Click to view available options
For Use With (Application):
Digital Pathology
Catalog No. For Use With (Application)
P4804080S Digital Pathology
1 options
Catalog No. P4804080S Supplier Epredia™ Supplier No. P480BF4080S
Add to Cart
Add to Cart
Epredia™ 3DHistech Pannoramic 480 whole-slide digital scanner is designed for labs that require reliable and automated high throughput while maintaining image quality and flexibility.
  • Largest-capacity, whole-slide scanner on the market: 480 standard-width slides
  • Can scan 15 x 15 mm slide area in 30 seconds (20x objective (40x optical equivalent), 0.25 μm/pixel, single layer)
  • Touch-enabled, easy-to-use software interface
  • Double-width slide capacity (optional)
  • Automated tissue detection and coverslip detection
  • Flexible Priority/STAT slide position allocation
  • Impressive Speed
  • Multilayer (Z-stack) and Extended Focus scanning (optional)
  • 3 individual objectives exchangeable automatically during scanning
  • 1D and 2D barcode reading and parsing - Research use only
Specifications
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.