Learn More
Epredia™ 3DHistech Pannoramic Desk II Digital Scanner, Brightfield to 55x Resolution
Catalog No. D2BF40S Shop All Epredia Products
Change view
Click to view available options
Resolution:
Brightfield to 55x
Catalog No. Resolution
D2BF40S Brightfield to 55x
1 options
Catalog No. D2BF40S Supplier Epredia™ Supplier No. D2BF40S
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic Desk II whole-slide digital scanner is an excellent choice for labs that need the highest quality imaging capabilities for their remote and frozen section workflow.
  • Double-wide slide capacity
  • Brightfield only scanning
  • 40x resolution by default, up to 55x
  • Manual slide loading, automatic previewing, barcode reading and scanning
  • Small footprint - Research use only
Specifications
Resolution Brightfield to 55x
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.