Learn More
Epredia™ 3DHistech Pannoramic MIDI Confocal Digital Scanner with Fluorescent Imaging, Plan-Apochromat 20x/0.8 NA, C-Apochromat w40x/1.2 NA Objectives
Catalog No. RCONFHA1025 Shop All Epredia Products
Change view
Click to view available options
20x, 40x
Catalog No.
RCONFHA1025 20x, 40x
1 options
Catalog No. RCONFHA1025 Supplier Epredia™ Supplier No. RCONFHAO0001025CEL
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic MIDI Confocal offers fast and high-quality confocal scanning for molecular pathology applications by combining confocal imaging with award-winning whole-slide scanning technology.
  • PANNORAMIC Confocal uses innovative structured illumination confocal imaging to overcome the limitations of spinning pinhole-disc techniques
  • This delivers the highest light efficiency with minimal bleaching and fastest scanning speed
  • Colocalized imaging is available with both fluorescent and brightfield illumination
  • Also, SW DDIC (Digital Differential Interference Contrast) is used to ensure low-contrast brightfield visualization - Research use only
Specifications
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.