Learn More
Epredia™ 3DHistech Pannoramic MIDI II Digital Scanner, Brightfield to 110x Resolution
Catalog No. M2BF55110S Shop All Epredia Products
Change view
Click to view available options
Resolution:
Brightfield to 110x
Catalog No. Resolution
M2BF55110S Brightfield to 110x
1 options
Catalog No. M2BF55110S Supplier Epredia™ Supplier No. M2BF55110S
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic MIDI II whole-slide digital scanner is a versatile, low-volume digital pathology solution for labs that need a compact, flexible scanner with BF and FL scanning capabilities of 12 slide batches.
  • Twelve-slide capacity and continuous loading with horizontal slide arrangement
  • Up to 110x brightfield resolution
  • Motorized objective changer
  • One high-quality monochrome camera with unique three-channel brightfield light source
  • Automatic slide loading, previewing, barcode reading and scanning - Research use only
Specifications
Resolution Brightfield to 110x
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.