Learn More
Epredia™ 3DHistech Pannoramic MIDI II Digital Scanner, Brightfield to 60x Resolution, Fluorescence to 60x Resolution - SOLA
Catalog No. M2BFFLSOLS Shop All Epredia Products
Change view
Click to view available options
Resolution:
Brightfield to 60x
Catalog No. Resolution
M2BFFLSOLS Brightfield to 60x
1 options
Catalog No. M2BFFLSOLS Supplier Epredia™ Supplier No. M2BF3060FLSOLS
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic MIDI II whole-slide digital scanner is a versatile, low-volume digital pathology solution for labs that need a compact, flexible scanner with BF and FL scanning capabilities of 12 slide batches.
  • Twelve-slide capacity and continuous loading with horizontal slide arrangement
  • Brightfield and up to nine fluorescent filter positions available for single and multiple band cubes
  • Up to 60x brightfield and 60x fluorescent resolution
  • Motorized objective changer
  • One high-quality monochrome camera is used for both brightfield and fluorescence with unique three-channel brightfield light source and Lumencor SOLA light source for fluorescent scanning
  • Automatic slide loading, previewing, barcode reading and scanning - Research use only
Specifications
Resolution Brightfield to 60x
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.