Learn More
Epredia™ 3DHistech Pannoramic MIDI II Digital Scanner, Brightfield to 80x Resolution, Fluorescence to 80x Resolution - SPECTRA
Catalog No. M3BFFLSPCS Shop All Epredia Products
Change view
Click to view available options
Resolution:
Brightfield to 80x
Catalog No. Resolution
M3BFFLSPCS Brightfield to 80x
1 options
Catalog No. M3BFFLSPCS Supplier Epredia™ Supplier No. M3BF204080FLSPCS
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic MIDI II whole-slide digital scanner is a versatile, low-volume digital pathology solution for labs that need a compact, flexible scanner with BF and FL scanning capabilities of 12 slide batches.
  • Twelve-slide capacity and continuous loading with horizontal slide arrangement
  • Brightfield and up to nine fluorescent filter positions available for single and multiple band cubes
  • Up to 80x brightfield and 80x fluorescent magnification
  • Motorized objective changer
  • One high-quality monochrome camera is used for both brightfield and fluorescence with unique three-channel brightfield light source and Lumencor SPECTRA light source for fluorescent scanning
  • Automatic slide loading, previewing, barcode reading and scanning
Specifications
Resolution Brightfield to 80x
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.