Learn More
Epredia™ 3DHistech Pannoramic SCAN II Digital Scanner, Brightfield to 110x Resolution, Fluorescence to 110x Resolution - SOLA
Catalog No. S2BF55110S Shop All Epredia Products
Change view
Click to view available options
110x, 110x
Catalog No.
S2BF55110S 110x, 110x
1 options
Catalog No. S2BF55110S Supplier Epredia™ Supplier No. S2BF55110S
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic SCAN II whole-slide digital scanner for labs that need BF and FL scanning, with continuous workflow loading capabilities of up to 150 slides at a time.
  • 150-slide capacity and continuous loading with vertical slide arrangement
  • Exceptional image quality for both brightfield and up to nine fluorescent filter positions available for single and multiple band cubes with advanced FISH scanning technique
  • Up to 110x brightfield and 110x fluorescent resolution by default
  • Motorized objective changer
  • One high-quality monochrome camera is used for both brightfield and fluorescence with unique three-channel brightfield light source and Lumencor SOLA light source for fluorescent scanning
  • Automatic slide loading, previewing, barcode reading and scanning
  • All-around system for high-volume slide scanning - Research use only
Specifications
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.