Learn More
Epredia™ 3DHistech Pannoramic SCAN II Digital Scanner, Brightfield to 110x Resolution
Catalog No. S2BF55110FL Shop All Epredia Products
Change view
Click to view available options
110x
Catalog No.
S2BF55110FL 110x
1 options
Catalog No. S2BF55110FL Supplier Epredia™ Supplier No. S2BF55110FLSOLS
Only null left
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech Pannoramic SCAN II whole-slide digital scanner for labs that need BF and FL scanning, with continuous workflow loading capabilities of up to 150 slides at a time.
  • 150-slide capacity and continuous loading with vertical slide arrangement
  • Exceptional image quality for brightfield
  • Up to 110x brightfield
  • Motorized objective changer
  • One high-quality monochrome camera with unique three-channel brightfield light source
  • Automatic slide loading, previewing, barcode reading and scanning
  • All-around system for high-volume slide scanning - Research use only
Specifications
Product Type Digital Scanner
For Use With (Application) Digital Pathology
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.