Learn More
Epredia™ 3DHistech PatternQuant Pattern Recognition Module - 3 Users
Catalog No. 91212 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Microarray Scanner
Catalog No. For Use With (Equipment)
91-212 Microarray Scanner
1 options
Catalog No. 91-212 Supplier Epredia™ Supplier No. 91212
Add to Cart
Add to Cart
Request a Quote
Epredia™ 3DHistech PatternQuant is a trainable pattern recognition module for tissue classification, tissue pre-segmentation and identification of several tissue structures.
  • Color- and pattern-based image analysis solutions are used to identify different tissue elements in stained tissue. Instead of the time-consuming manual annotation, automated detection and annotation of a region of interest is available with a quick training on relevant tissue samples.
  • PatternQuant is a trainable pattern recognition module for tissue classification, tissue pre-segmentation and identification of several tissue structures. The artificial intelligence-based algorithm is able to learn and classify various tissue types based on their texture patterns and color features. This module can analyze fluorescent and brightfield slides.
  • License for 3 users - Research use only
Specifications
Product Type Pattern Recognition Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.