Learn More
Epredia™ 3DHistech PatternQuant Plus Pattern Recognition Module - 3 Users
Catalog No. 91072 Shop All Epredia Products
Change view
Click to view available options
3 Users
Catalog No.
91-072 3 Users
1 options
Catalog No. 91-072 Supplier Epredia™ Supplier No. 91072
Add to Cart
Add to Cart
Epredia™ 3DHistech PatternQuant Plus is an improved alternative to PatternQuant for more challenging samples and for complex tissue segmentation projects when thorough training using deep learning on multiple slides is needed.
  • Color- and pattern-based image analysis solutions are used to identify different tissue elements in stained tissue. Instead of the time-consuming manual annotation, automated detection and annotation of a region of interest is available with a quick training on relevant tissue samples.
  • PatternQuant Plus is an improved alternative to PatternQuant for more challenging samples (low visual difference between the staining of tissue types within the sample) and for complex tissue segmentation projects when thorough training using deep learning on multiple slides is needed. This module can analyze fluorescent and brightfield slides.
  • License for 3 users - Research use only
Specifications
Product Type Pattern Recognition Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner
For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.