Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ 3DHistech ScriptQuant Image Analysis Scripts Module - 3 Users

Catalog No. 91262 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Microarray Scanner
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
91-262 Microarray Scanner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-262 Supplier Epredia™ Supplier No. 91262
Add to Cart
Add to Cart
Request a Quote

Epredia™ 3DHistech ScriptQuant image analysis scripts module provides a shell for user-created image analysis scripts in Python for maximum flexibility for the research user, enabling them to run their own image analysis algorithms within the QuantCenter framework.

A unique tool for maximum flexibility.

ScriptQuant is a brand-new image analysis module for QuantCenter. It provides a shell for user-created image analysis scripts in Python for maximum flexibility for the research user, enabling them to run their own image analysis algorithms within the QuantCenter framework.

Key Features

  • Arbitrary script editor (VS Code by Microsoft is recommended)
  • The user-created script can be saved into a scenario and handled like one
  • Fast visual feedback: write your code and see the results immediately
  • ScriptQuant can be extended with external libraries (e.g. OpenCV, TensorFlow)
  • Well-documented API and use case examples
  • ScriptQuant may provide a gateway for 3rd party AI developers
  • Quantification results can be fed back into the 3DHISTECH infrastructure:
    • Storage of quantification results (polygons, objects, regions, measured values)
    • Visualization in SlideViewer as an overlay
    • Statistics and Data visualization
  • License for 3 users - Research use only

Specifications

Product Type Image Analysis Script Module
For Use With (Application) Digital Pathology
For Use With (Equipment) Microarray Scanner

For research and education use only, not for use in diagnostic procedures in the U.S. This product has not been approved or cleared as a medical device by the U.S. Food and Drug Administration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.